Learn More
Medchemexpress LLC DSP-1053 | 1176326-76-3 | 99.8% | 502.44 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY111419
Encompass_Preferred
DSP-1053, a benzylpiperidine derivative, is a potent Serotonin Transporter (SERT) inhibitor with a Ki of 1.02 nM. It also exhibits partial 5-HT1A receptor agonistic activity with a Ki of 5.05 nM and possesses antidepressant activity.
  • Potent Serotonin Transporter (SERT) inhibitor
  • Partial 5-HT1A receptor agonistic activity
  • Antidepressant activity
Catalog No. 50-003-46293