Learn More
Bioworld DSP, 1 g
Supplier:  Bioworld 218200051
Encompass_Preferred
DSP, Dithiobis(succinimidyl propionate)Primary amine reactiveThiol cleavableReferences: 1.) Lomant, A.J. and Fairbanks, G. (1976) J. Mol. Biol. 104, 243-261. 2.) Tavers, R.C., et al. (1982) J. Biol. Chem. 257, 10708-10714.M.W. = 404.4
Catalog No. 50-199-280
 Restricted Item