Learn More
Medchemexpress LLC Dspe-biotin | 133695-76-8 | 97.5% | 974.36 | C51H96N3O10PS | 25 MG

Supplier: Medchemexpress LLC HYW44100025MG
DSPE-Biotin is a biotin-functionalized phospholipid designed to introduce biotin into lipid-based systems such as liposomes and micelles for streptavidin/avidin-mediated capture, labeling, or immobilization in biochemical and biophysical research applications.
- Biotin functional group enables strong streptavidin/avidin binding.
- Compatible with incorporation into liposomes, micelles, and other lipid assemblies.
- High purity (97.5%) supports reproducible experimental results.
- Solid, off-white to light yellow appearance for easy handling.
- Soluble in DMSO (≈1.3 mg/mL) with sonication and warming to 60°C.
- Store protected from light at -20°C; in solvent, store at -80°C for up to 6 months.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.