Learn More
Medchemexpress LLC Dspe-m-peg-nhs | MFCD03098978 | 99.4% | 3,400 (average) | 5 MG
SDP
Supplier:  Medchemexpress LLC HYW441015A5MG
Encompass_Preferred
DSPE-m-PEG-NHS (average MW 3,400) is a pegylated phospholipid with an NHS-activated ester used to introduce PEG chains onto lipid bilayers and to covalently attach amine-containing ligands. It is supplied as a powder for formulation of liposomes and lipid nanoparticles, including applications in nucleic acid vaccine delivery, and is characterized for use in reproducible nanoparticle preparation.
  • Pegylated phospholipid with an NHS-activated ester for amine coupling.
  • Suitable for liposome and lipid nanoparticle formulation for targeted delivery.
  • Average molecular weight 3,400 Da provides steric stabilization.
  • High purity for reproducible formulation performance.
  • Powder form with recommended freezer storage for long-term stability.
Catalog No. 50-002-81101