Learn More
Medchemexpress LLC DSPE-Mal | 1360858-99-6 | 95.0% | 899.18 g/mol | C48H87N2O11P | 50 MG
SDP
Supplier:  Medchemexpress LLC HYW44100750MG
Encompass_Preferred
DSPE-Mal is a maleimide-functional phospholipid used to introduce thiol-reactive maleimide groups into lipid assemblies and liposomes, enabling covalent coupling of thiol-containing ligands for targeted nanocarrier and surface functionalization applications.
  • Maleimide-functionalized phospholipid for thiol-specific conjugation.
  • Facilitates stable covalent coupling of thiol-containing ligands to lipid bilayers.
  • Amphiphilic structure supports self-assembly into liposomes and vesicles.
  • Useful for targeted drug delivery and nanocarrier surface modification.
  • Offered in small research-scale quantities for formulation studies.
Catalog No. 50-002-81446