Learn More
TARGETMOL CHEMICALS INC DSPE-PEG 2000 100MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T74229100MG
Encompass_Preferred
Also available in 5 mg, 10 mg, 25 mg, 50 mg, 500 mg and bulk. Please contact Fisher for quotes. DSPE-PEG 2000 is a non-toxic and biodegradable amphiphilic copolymer that can be used to form micelles to deliver nanoparticles.
Catalog No. 50-287-6271