Learn More
Medchemexpress LLC DSPE-PEG 2000 | 892144-24-0 | 99.37% | (C2H4O)nC42H82NO10P | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY142979
Encompass_Preferred
DSPE-PEG 2000 is a PEG polymer containing DSPE and amine end groups. It can be used to form micelles as nanoparticles for drug delivery.
  • Powder: -20°C for 3 years.
  • In solvent: -80°C for 6 months, -20°C for 1 month.
Catalog No. 50-003-54736