Learn More
Medchemexpress LLC DSPE-PEG 2000 | 892144-24-0 | 99.37% | (C2H4O)nC42H82NO10P | 250 MG

Supplier: Medchemexpress LLC 250MGHY142979
DSPE-PEG 2000 is a PEG polymer that contains DSPE and amine end groups. It can be utilized to form micelles as nanoparticles for drug delivery and is intended for research use only.
- Contains DSPE and amine end groups
- Can be used to form micelles as nanoparticles for drug delivery
- For research use only
- Solid appearance
- White to off-white color
- Powder storage: -20°C for 3 years
- In solvent storage: -80°C for 6 months, -20°C for 1 month
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.