Learn More
Medchemexpress LLC Dspe-peg 2000 | 892144-24-0 | 99.37% | (C2H4O)nC42H82NO10P | 500 MG
SDP
Supplier:  Medchemexpress LLC HY142979500MG
Encompass_Preferred
DSPE-PEG 2000 is a PEG polymer that incorporates DSPE and amine end groups. This compound can be effectively utilized for forming micelles, which are critical nanoparticles for advanced drug delivery systems.
  • Forms micelles as nanoparticles
  • Suitable for drug delivery applications
  • Contains DSPE and amine end groups
Catalog No. 50-003-56682