Learn More
Medchemexpress LLC Dspe-peg-biotin | 98.4% | 3400 Da (average) | 100 MG
SDP
Supplier:  Medchemexpress LLC HYW440886100MG
Encompass_Preferred
DSPE-PEG-Biotin (average MW 3400) is a phospholipid-polyethylene glycol conjugate designed for biotinylation and incorporation into lipid assemblies. Its amphiphilic character supports use in liposomes, micelles, and targeted drug delivery research, and it is supplied as a solid for laboratory handling.
  • Facilitates biotinylation of molecules.
  • Amphiphilic; integrates into lipid membranes and liposomes.
  • Average molecular weight 3400 Da.
  • Provided as a white solid for convenient handling.
  • High purity (≈98.4%) suitable for research applications.
  • Powder stable at -20°C; in solvent store at -80°C for longer-term storage.
Catalog No. 50-002-81463