Learn More
TARGETMOL CHEMICALS INC DSPE-PEG-Biotin MW 2000 25MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T1785025MG
Encompass_Preferred
Also available in 2 mg, 5 mg, 10 mg, 50 mg and bulk. Please contact Fisher for quotes. DSPE-PEG-Biotin (MW 2000) is a PEG-based PROTAC linker for PROTAC synthesis. Purity 95%
Catalog No. 50-287-6033