Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC DSPE-PEG-CHO (MW 2000) | 89.0% | 2000 (Average) | 5 MG
SDP

Supplier:  Medchemexpress LLC HY176509A5MG

Encompass_Preferred

DSPE-PEG-CHO (MW 2000) is a chemical compound with an average molecular weight of 2000. It appears as a white to off-white solid and has been tested to comply with given specifications. This product is intended for research use only.

  • Appearance: White to off-white solid
  • 1H NMR Spectrum consistent with structure
  • Purity (HPLC): 89.04%
  • Storage as powder: -20°C for 3 years
  • Storage in solvent: -80°C for 6 months, -20°C for 1 month

Catalog No. 50-003-87237


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.