Learn More
Medchemexpress LLC DSPE-PEG-COOH | 100.0% | 2000 (average) | 50 MG
SDP
Supplier:  Medchemexpress LLC HYW59146150MG
Encompass_Preferred
DSPE-PEG-COOH (MW 2000) is a PEGylated phospholipid with a terminal carboxylic acid used to prepare liposomes and lipid nanoparticles and to enable covalent surface conjugation via amide bond formation. Supplied as a white to off-white solid for research use.
  • Provides carboxylic acid terminal for amide coupling with primary amines.
  • Average PEG molecular weight of 2000 Da for predictable steric stabilization.
  • Suitable for liposome and lipid nanoparticle formulation and surface functionalization.
  • White to off-white solid supplied in laboratory quantities for research applications.
  • High purity appropriate for experimental use.
  • Store sealed, away from moisture, at low temperatures for stability.
Catalog No. 50-002-84778