Learn More
Medchemexpress LLC Dspe-peg-cooh | 98.1% | 5000 (Average) | 50 MG
SDP
Supplier:  Medchemexpress LLC HY16696850MG
Encompass_Preferred
DSPE-PEG-COOH (MW 5000) is a PEG lipid that can improve the delivery efficiency and tissue specificity of poorly soluble drugs. It can be used in drug delivery research.
  • Improves delivery efficiency
  • Enhances tissue specificity
  • Suitable for drug delivery research
  • PEG lipid
Catalog No. 50-003-90930