Learn More
Medchemexpress LLC DSPE-PEG-DBCO | 99.9% | 2000 (average) | 25 MG
SDP
Supplier:  Medchemexpress LLC HYW44083525MG
Encompass_Preferred
DSPE-PEG-DBCO (MW 2000) is a PEGylated phospholipid conjugate bearing a dibenzocyclooctyne (DBCO) group. It serves as a click-chemistry-capable lipid for incorporation into lipid nanoparticles or micelles and for rapid, copper-free conjugation to azide-containing molecules via strain-promoted alkyne-azide cycloaddition (SPAAC). Typical uses include nanoparticle formulation, surface PEGylation, and bioconjugation.
  • PEGylated phospholipid with average molecular weight 2000.
  • DBCO functional group enables copper-free SPAAC with azides.
  • Suitable for lipid nanoparticle and micelle formulation.
  • Facilitates bioconjugation and surface modification.
  • High purity (reported 99.9%) for research applications.
Catalog No. 50-002-81460