Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC DSPE-PEG-DBCO sodium | 2052955-83-4 | 95.0% | 5000 (average) | 1 MG
SDP

Supplier:  Medchemexpress LLC HYW4408941MG

Encompass_Preferred

DSPE-PEG-DBCO sodium (MW 5000) is a PEGylated phospholipid that combines a DSPE lipid anchor with a dibenzocyclooctyne (DBCO) reactive handle, designed for incorporation into lipid formulations and for strain-promoted alkyne-azide cycloaddition (SPAAC) bioconjugation in drug delivery research.

  • Pegylated lipid with DBCO functionality for copper-free click chemistry.
  • Sodium salt form supplied as a white to yellow solid.
  • Average molecular weight approximately 5000 daltons.
  • Purity approximately 95% by supplier analysis.
  • Enables liposome and nanoparticle surface functionalization.
  • Store at -20°C and protect from light; follow solvent storage guidance for long-term stability.

Catalog No. 50-002-81172


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.