Learn More
Medchemexpress LLC DSPE-PEG-endo-BCN | 92.1% | 5000 (Average) | 1 MG
SDP
Supplier:  Medchemexpress LLC HY1668711MG
Encompass_Preferred
DSPE-PEG-endo-BCN (MW 5000) is a PEG lipid that can improve the delivery efficiency and tissue specificity of poorly soluble drugs. It can be used in drug delivery research.
  • Improves delivery efficiency of poorly soluble drugs
  • Enhances tissue specificity
  • Suitable for drug delivery research
Catalog No. 50-003-92340