Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC DSPE-PEG-FITC | 1000788-53-3 | 10000 Da | 5 MG
SDP

Supplier:  Medchemexpress LLC HYW440914B5MG

Encompass_Preferred

DSPE-PEG-FITC is a fluorescein-labeled PEGylated phospholipid designed for incorporation into liposomes, micelles, and other lipid-based nanoparticles to provide a green fluorescent tag for tracking and imaging in research applications.

  • Average molecular weight 10,000 Da.
  • Fluorescein (FITC) label provides green fluorescence for imaging.
  • Suitable for liposome and nanoparticle labeling and tracking.
  • Appearance: yellow to orange solid.
  • Soluble in DMSO; use ultrasonic and warming up to 60°C to aid dissolution.
  • In solvent stability: store at -80°C up to 6 months or -20°C up to 1 month; protect from light.
  • Available in small research sizes such as 5 mg.
  • Recommended in vivo formulation: 10% DMSO in 90% corn oil for administration.

Catalog No. 50-002-81265


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.