Learn More
Medchemexpress LLC Dspe-peg-fitc | 1000788-53-3 | >95.0% | 10000 Da (average) | 10 MG
SDP
Supplier:  Medchemexpress LLC HYW440914B10MG
Encompass_Preferred
DSPE-PEG-FITC (MW 10000) is a PEGylated phospholipid conjugated to fluorescein isothiocyanate, designed as an amphiphilic fluorescent lipid for incorporation into liposomes, nanoparticles, or for micelle formation in aqueous solutions. The product is supplied as a solid with an average PEG molecular weight of 10,000 and is used for fluorescent labeling and tracking in drug delivery and nanocarrier research.
  • Amphiphilic PEGylated phospholipid suitable for liposome and nanoparticle labeling.
  • Fluorescent FITC tag enables visualization and tracking in imaging and biodistribution studies.
  • Average PEG molecular weight 10,000 provides steric stabilization and extended circulation properties.
  • Forms micelles in aqueous solution for easy incorporation into nanocarriers.
  • Solid appearance with recommended storage at -20°C; protect from light (in solvent: -80°C for 6 months, -20°C for 1 month).
Catalog No. 50-002-80991