Learn More
Medchemexpress LLC DSPE-PEG-FITC | 1000788-53-3 | 98.1% | 5000 Da (average) | 10 MG
SDP
Supplier:  Medchemexpress LLC HYW44091710MG
Encompass_Preferred
DSPE-PEG-FITC (MW 5000) is a fluorescein-conjugated polyethylene glycol (PEG) lipid amphiphile designed for fluorescent labeling of liposomes, nanoparticles, and biological samples. It combines a DSPE lipid anchor with an FITC-labeled PEG chain to provide stable fluorescence for imaging and tracking applications. Supplied as a light yellow solid, handle and store per manufacturer guidance.
  • Provides fluorescein (FITC) fluorescence for detection and imaging.
  • Average PEG molecular weight ≈ 5,000 Da.
  • Amphiphilic PEG-lipid suitable for incorporation into liposomes and micelles.
  • High purity suitable for research use.
  • Light yellow solid physical form for easy handling.
  • Storage recommendations available for powder and solutions.
Catalog No. 50-002-81363