Learn More
Medchemexpress LLC Dspe-peg-fitc | 600 g/mol (average) | 5 MG
SDP
Supplier:  Medchemexpress LLC HYW440914C5MG
Encompass_Preferred
DSPE-PEG-FITC (MW 600) is a fluorescein-labeled PEGylated phospholipid used as a fluorescent lipid probe for preparation of liposomes and nanoparticles, surface labeling, and tracking in research applications. The material is supplied as a light yellow to yellow solid and is intended for laboratory research; storage is recommended at 4°C protected from light, and in solvent at -80°C for up to 6 months or at -20°C for up to 1 month.
  • Fluorescent labeling for liposomes and nanoparticles.
  • Amphiphilic PEGylated phospholipid enabling micelle and liposome formation.
  • Average molecular weight 600 for consistent labeling performance.
  • Light yellow solid for convenient storage and handling.
  • Stable when stored protected from light; solvent storage at -80°C (6 months) or -20°C (1 month).
Catalog No. 50-002-80992