Learn More
Medchemexpress LLC Dspe-peg-fitc | 95.0% | 600 Da (average) | 10 MG

Supplier: Medchemexpress LLC HYW440914C10MG
Fluorescein-labeled PEGylated phospholipid supplied as a powder with an average molecular weight of approximately 600 Da. This reagent-grade fluorescent lipid conjugate is designed for incorporation into liposomes and lipid nanoparticles to enable fluorescent labeling, tracking, and imaging in laboratory research. It is supplied for research use only and can form micelles in aqueous solution.
- Fluorescent pegylated phospholipid for liposome and nanoparticle labeling.
- Average molecular weight 600 Da (average).
- Reagent grade powder suitable for research use.
- Forms micelles in aqueous solution.
- Purity 95.0% per certificate of analysis.
- Store protected from light at 4°C; in solvent store at -80°C for up to 6 months.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.