Learn More
Medchemexpress LLC DSPE-PEG-Folate | 1236288-25-7 | 95.7% | 3400.00 | C65H114N11O16P | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1440095MG
Encompass_Preferred
DSPE-PEG-Folate is a pegylated phospholipid bearing a terminal folate group that enables folate-receptor mediated targeting. It integrates into lipid bilayers or forms micelles for use in liposome and nanoparticle drug-delivery research and targeting studies.
  • Enables folate-mediated targeting to folate receptor-expressing cells.
  • Integrates into lipid bilayers and forms micelles for nanoparticle and liposome formulation.
  • Terminal folate provides a handle for receptor-specific delivery.
  • High purity suitable for research applications (reported ~95.7%).
  • Available in small research pack sizes for formulation screening.
Catalog No. 50-002-21559