Learn More
Medchemexpress LLC DSPE-PEG-maleimide | 00-00-0 | 99.5% | 10MG

Supplier: Medchemexpress LLC HY14074010MG
DSPE-PEG-maleimide (MW 5000) is a pegylated phospholipid that combines a DSPE lipid anchor with a PEG spacer terminated in a maleimide group for selective thiol conjugation. Supplied as a solid powder with high purity, it is designed for incorporation into liposomes and lipid nanoparticles and for attaching thiol-containing ligands and peptides to particle surfaces.
- Molecular weight ~5,000 for predictable spacer length.
- Maleimide functional group enables selective thiol conjugation.
- DSPE lipid anchor allows stable insertion into lipid bilayers.
- PEG spacer increases circulation time and stability of formulations.
- High purity (~99.5%) suitable for research and formulation work.
- Powder form with recommended storage at -20°C for long-term stability.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.