Learn More
Medchemexpress LLC DSPE-PEG-Maleimide | 1612754-08-1 | 99.9% | 2000 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY166648
Encompass_Preferred
DSPE-PEG-Maleimide is a functional lipid component and a thiol-reactive crosslinker. It undergoes Michael addition with the thiol groups of thiolated or cyclic RGD peptides to form stable thioether bonds and DSPE-PEG (2000)-RGD, making it suitable for research on drug delivery. In vitro, DSPE-PEG-Maleimide successfully conjugates with one thiolated RGD peptide via Michael addition at 4°C, pH 8 to form DSPE-PEG(2000)-RGD.
  • Functional lipid component and thiol-reactive crosslinker
  • Forms stable thioether bonds via Michael addition with thiol groups
  • Applicable for research on drug delivery
  • Conjugates with thiolated RGD peptides
Catalog No. 50-003-95249