Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC DSPE-PEG-Maleimide ammonium | 99.0% | 2000 | 1 MG
SDP

Supplier:  Medchemexpress LLC HY166648A1MG

Encompass_Preferred

DSPE-PEG-Maleimide (ammonium) (MW 2000) contains DSPE phospholipids and maleimide, used to prepare nanostructured lipid carriers and in drug delivery research.

  • Appearance: Solid
  • Color: White to off-white
  • Shipping: Room temperature in continental US; may vary elsewhere
  • Storage: -20°C, sealed storage, away from moisture. In solvent: -80°C, 6 months; -20°C, 1 month (sealed storage, away from moisture)

Catalog No. 50-003-91721


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.