Learn More
Medchemexpress LLC DSPE-PEG-Maleimide, MW 5000 | 99.8% | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY140740
Encompass_Preferred
DSPE-PEG-Maleimide, with an average molecular weight of 5000, is a linker lipid composed of DSPE phospholipid and Maleimide. This compound is primarily utilized in the synthesis of various liposome types, including HELDC, sterically stabilized, and immunoliposomes.
  • Can be coupled with thiol-p5314-29 peptide
  • Used as a linker lipid in immunoliposome synthesis
  • Sterically stabilized liposomes containing this compound can be coupled with thiolated monoclonal antibody OX26 (OX26-SH)
  • HELDC liposomes prepared with 3% DSPE-PEG-Maleimide show stable particle size over two weeks, with an average diameter of 120 nm
Catalog No. 50-003-57092