Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Dspe-peg-mannose | 1829524-73-3 | 90.0% | 2000 Da (average) | (C2H4O)nC57H102N3O16PS | 10 MG
SDP

Supplier:  Medchemexpress LLC HY16837410MG

Encompass_Preferred

DSPE-PEG(2000)-Mannose is a mannose-terminated DSPE-PEG lipid conjugate used to prepare mannose-functionalized liposomes and lipid nanoparticles for receptor-mediated targeting. It is intended for research use to enhance targeting of antigen-presenting cells and to improve vaccine or drug delivery performance.

  • Lipid-PEG conjugate with a terminal mannose for mannose-receptor targeting.
  • Average molecular weight ~2000 Da.
  • Purity 90.0%.
  • Appearance: white to off-white solid.
  • Storage: powder -20°C (3 years); in solvent -80°C (6 months), -20°C (1 month).
  • Suitable for constructing mannose-functionalized liposomes and lipid nanoparticles for targeting applications.
  • CAS number 1829524-73-3.

Catalog No. 50-002-89432


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.