Learn More
Broadpharm DSPE-PEG-Mannose, MW 2000, 50 mg, PEG Lipid, Solid, Purity 90%, Storage -20°C, Shipping Ambient Temperature
Supplier:  Broadpharm BP4040750MG
Encompass
DSPE-PEG-Mannose, MW 2000 is a specialized polyethylene glycol (PEG) lipid, specifically engineered for advanced drug delivery systems, particularly in lipid nanoparticle (LNP) formulations. This innovative conjugate includes a DSPE phospholipid and a mannose sugar residue, facilitating targeted delivery of therapeutics and enhancing the stability of formulations. The unique properties of DSPE allow for micelle formation, essential for lipid nanoparticle construction. Additionally, the PEG component significantly increases water solubility and prolongs circulation time in vivo, making it advantageous for delivering mRNA vaccines and other targeted therapies.
  • Facilitates targeted drug delivery systems
  • Enhances stability for advanced therapies
  • Allows for micelle formation, crucial for lipid nanoparticles
  • Increases water solubility and circulation time
  • Suitable for targeted liposomal delivery systems
Catalog No. NC2480925