Learn More
Medchemexpress LLC Dspe-peg-nhs | 98.0% | 2000 (average) | 10 MG

Supplier: Medchemexpress LLC HYW44101410MG
DSPE-PEG-NHS, MW 2000 is a PEG-modified phospholipid with an N-hydroxysuccinimide (NHS) activated ester for conjugation to primary amines. It is used to modify liposome surfaces and other lipid-based nanoparticles, providing a lipid anchor with a PEG spacer to improve colloidal stability and enable surface functionalization for drug delivery research.
- NHS-activated ester for efficient amine conjugation.
- DSPE lipid anchor for stable membrane insertion.
- PEG spacer (average molecular weight 2000) provides steric stabilization and reduced opsonization.
- High purity (~98.0%) suitable for research applications.
- Solid, white to off-white form; store at -20 °C sealed away from moisture.
- Available in small vials (e.g., 10 MG) for lab-scale work.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.