Learn More
Medchemexpress LLC Dspe-peg-oh (mw 2000) | 99.9% | 50 MG
SDP
Supplier:  Medchemexpress LLC HY14074150MG
Encompass_Preferred
DSPE-PEG-OH (MW 2000) is a hydroxyl-terminated PEGylated phospholipid. Its hydrophobic tail enables encapsulation and aggregation of other hydrophobic drugs, and its hydroxyl terminus is suitable for further reactions. This product can be used to prepare liposomes or lipid nanoparticles.
  • Hydroxyl-terminated PEGylated phospholipid
  • Hydrophobic tail for encapsulation and aggregation of hydrophobic drugs
  • Hydroxyl terminus suitable for further reactions
  • Can be used to prepare liposomes or lipid nanoparticles
Catalog No. 50-003-54944