Learn More
Medchemexpress LLC Dspe-PEG-SH | 1462982-18-8 | 90.0% | C44H86NO10PS | 1 G
SDP
Supplier:  Medchemexpress LLC HYW4408951G
Encompass_Preferred
DSPE-PEG-SH (MW 1000) is a thiol-functionalized pegylated phospholipid used to introduce reactive thiol groups onto lipid membranes and biomolecules. Supplied as an average MW ~1000 PEG-lipid, it is used for liposome and lipid nanoparticle preparation, surface modification, and thiol-maleimide conjugation to attach ligands or drugs.
Catalog No. 50-002-81129