Learn More
Medchemexpress LLC DSPE-PEG-SH | 1462982-18-8 | 99.4% | 2000 Da (average) | C44H86NO10PS | 50 MG
SDP
Supplier:  Medchemexpress LLC HYW44089650MG
Encompass_Preferred
DSPE-PEG-SH is a thiol-terminated pegylated phospholipid (average MW 2000) designed for incorporation into lipid bilayers and lipid nanoparticle formulations. The terminal -SH group enables covalent surface functionalization and bioconjugation for targeted delivery or conjugate assembly.
  • Thiol-terminated PEG2000 conjugated to distearoyl phosphoethanolamine.
  • Amphipathic lipid integrates into lipid bilayers and nanoparticles.
  • Reactive thiol group for covalent conjugation with maleimide.
  • Average molecular weight 2000 Da.
  • High purity (≈99.4%).
  • White to off-white solid with defined solubility in water and DMSO when warmed.
  • Powder stable at -20°C; in solution store at -80°C for long-term stability.
Catalog No. 50-002-80501