Learn More
Medchemexpress LLC DSPE-PEG-SH | 90.0% | 10 MG
SDP
Supplier:  Medchemexpress LLC HYW44089510MG
Encompass_Preferred
DSPE-PEG-SH (MW 1000) is a thiol-terminated pegylated phospholipid consisting of a DSPE hydrophobic anchor and a polyethylene glycol chain with an average molecular weight of 1000. It self-assembles into lipid bilayers and is commonly used to functionalize liposomes and lipid nanoparticles. The terminal thiol enables site-specific conjugation for attaching ligands, peptides, or probes, while the PEG chain imparts steric stabilization and improved circulation properties.
  • Thiol terminal for site-specific conjugation.
  • DSPE anchor for stable membrane incorporation.
  • Pegylated chain for steric stabilization and reduced opsonization.
  • Average molecular weight 1000 for short PEG spacer properties.
  • Suitable for liposome and lipid nanoparticle functionalization.
Catalog No. 50-002-81641