Learn More
Nanocs DSPE PEG Thiol, DSPE-PEG-SH, 3400 Da, 100mg
Supplier:  Nanocs PG2DSTH3K
Encompass
1,2-Distearoyl-sn-Glycero-3-Phosphoethanolamine (DSPE) conjugated Polyethylene Glycol, DSPE PEG is a phospholipid PEG conjugate which has both hydrophilicity and hydrophobility. Pegylated phospholipids are excellent liposome formation materials that can be used for drug delivery, gene transfection and vaccine delivery as well. Pegylation of phospholipids significantly improves the blood circulation time and stability for encapsulated drugs. These materials can also be used for targeted drug delivery by modifying their surfaces with targeting ligands such as antibodies, peptides.
Catalog No. NC3793275