Learn More
Medchemexpress LLC DSPE-PEG4-DBCO | 2112738-14-2 | 95.0% | C71H116N3O15P | 100 MG
SDP
Supplier:  Medchemexpress LLC HY140955100MG
Encompass_Preferred
DSPE-PEG4-DBCO is a PEG-based PROTAC linker employed in the synthesis of PROTACs. It serves as a click chemistry reagent, incorporating a DBCO group that facilitates strain-promoted alkyne-azide cycloaddition (SPAAC) with azide-containing molecules. This mechanism leverages the intracellular ubiquitin-proteasome system for the selective degradation of target proteins.
  • PEG-based linker
  • Utilized in PROTAC synthesis
  • Functions as a click chemistry reagent
  • Contains a DBCO group
  • Facilitates strain-promoted alkyne-azide cycloaddition (SPAAC)
  • Enables SPAAC with azide-containing molecules
  • Leverages the ubiquitin-proteasome system
  • Achieves selective protein degradation
Catalog No. 50-003-55129