Learn More
Medchemexpress LLC DSPE-PEG6-Mal | 95.0% | 1163.50 | 25 MG

Supplier: Medchemexpress LLC 25MGHY138395
DSPE-PEG6-Mal is a chemical compound identified by the molecular formula C60H111N2O17P and a molecular weight of 1163.50. It appears as a white to off-white solid.
- Appears as a white to off-white solid
- 1H NMR spectrum consistent with its structure
- Purity of 95.0% confirmed by NMR
- Store at -20°C in sealed container, away from moisture
- If in solvent, store at -80°C for up to 6 months or -20°C for 1 month in sealed container, away from moisture
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.