Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC DSPE-succinic acid | 248253-94-3 | 98.0% | 848.14 g/mol | C45H86NO11P | 10 MG
SDP

Supplier:  Medchemexpress LLC HYW44100210MG

Encompass_Preferred

DSPE-succinic acid is a phospholipid functionalized with a terminal succinic (carboxylic acid) moiety. The carboxylic acid is reactive toward amines, enabling stable amide conjugation for lipid functionalization and for preparing liposomes or nanoparticle drug carriers.

  • Reactive carboxylic acid headgroup for amide coupling.
  • Suitable for liposome and nanoparticle formulation.
  • High purity (98.0%).
  • Molecular weight 848.14 g/mol.
  • Soluble in DMF (3.33 mg/mL) with sonication and warming to 60°C.
  • Store powder at -20°C; in solvent store at -80°C up to 6 months.

Catalog No. 50-002-81719


May include imposed supplier surcharges.
Only null left
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.