Learn More
Medchemexpress LLC Dspe-thiol | 144735-82-0 | MFCD29079392 | 90.0% | 836.19 g/mol | C44H86NO9PS | 1 MG
SDP
Supplier:  Medchemexpress LLC HYW4410041MG
Encompass_Preferred
DSPE-Thiol is a thiol-capped distearoylphosphatidylethanolamine used to introduce reactive sulfhydryl groups into lipid assemblies. The thiol headgroup enables selective covalent coupling via thiol-maleimide chemistry and can be used to create functionalized liposomes or phospholipid dimers.
  • Thiol headgroup enables selective reaction with maleimide reagents.
  • Facilitates preparation of phospholipid dimers and lipid conjugates.
  • Useful for liposome functionalization and surface modification.
  • Supplied as a research-grade phospholipid with documented purity and molecular data.
Catalog No. 50-002-81012