Learn More
Medchemexpress LLC DSPE-Thiol | 144735-82-0 | MFCD29079392 | 90.0% | 836.19 g·mol⁻¹ | C44H86NO9PS | 5 MG
SDP
Supplier:  Medchemexpress LLC HYW4410045MG
Encompass_Preferred
A thiol-capped distearoyl phosphatidylethanolamine (DSPE) phospholipid for thiol-based conjugation and lipid surface modification. The terminal thiol selectively reacts with maleimide and other thiol-reactive groups, enabling preparation of phospholipid dimers, surface functionalization of liposomes or lipid nanoparticles, and formation of stable thiol-linked conjugates. Supplied as a solid with reported solubility information for DMF and typical purity at 90.0%.
  • Thiol-functionalized head group for selective thiol-maleimide conjugation.
  • Suitable for liposome surface modification and nanoparticle functionalization.
  • Enables preparation of phospholipid dimers and thiol-linked conjugates.
  • Soluble in DMF with sonication and gentle heating to aid dissolution.
  • Typical purity 90.0% and molecular weight 836.19 g·mol⁻¹.
Catalog No. 50-002-81114