Learn More
Medchemexpress LLC HY-110120 10mg Medchemexpress, DSR-6434 CAS:1059070-10-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY11012010MG
Encompass_Preferred
Medchemexpress, HY-110120 10mg DSR-6434 CAS:1059070-10-8 DSR-6434 is a potent and selective Toll-like receptor 7 (TLR7) agonist, with EC50s of 7.2 nM and 4.6 nM for human and mice TLR7, respectively. DSR-6434 has a strong antitumor effect. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8751