Learn More
Medchemexpress LLC HY-W021879 50mg , DSRM-3716 CAS:58142-99-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYW02187950MG
Encompass_Preferred
HY-W021879 50mg DSRM-3716 CAS: 58142-99-7 DSRM-3716 (5-Iodoisoquinoline) is a potent and selective SARM1 NADase inhibitor with an IC50 of 75 nM. DSRM-3716 is selective against other NAD+-processing enzymes, receptors, and transporters. DSRM-3716 provides robust axon protection. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0338