Learn More
Vector Laboratories Scicons Anti-dsRNA ELISA Kit, J2 Based
SDP
Supplier:  Vector Laboratories 10613005
Encompass
Double-stranded RNA (dsRNA) ELISA kit containing the following reagents for 500 tests (5x96 wells):
• 1 vial of coating antibody (store at -20 °C)
• 1 vial of Poly(I:C) dsRNA a positive control (store at -20 °C)
• 1 vial of dsRNA-specific detecting antibody (in RPMI + 5% FBS, store at +4 °C or, preferably, at -20 °C)
• 1 vial of HRP-conjugated F(ab’)2 Fragment of goat-anti mouse secondary antibody (store at +4 °C or at -20 °C)
• 1 vial of TMB substrate solution (store at +4°C, keep in dark)
Catalog No. NC1977292