Learn More
Kyfora Bio DSTAP 500 MG
Supplier:  Kyfora Bio 14486500
Encompass_Preferred
DSTAP chloride (1,2-distearoyl-3-trimethylammonium-propane chloride) is a cationic lipid and a with a variety of potential applications, including transfection and drug delivery.
Catalog No. 50-255-9838