Learn More
Perkin Elmer US LLC D-Swafer Dean's Switch
Supplier:  Perkin Elmer US LLC N9306251
Encompass_Preferred
Swafer micro-channel wafer technology is an innovative and user-friendly approach for flowswitching and splitting applications - it delivers unparalleled hardware and application flexibility, expanding the capabilities of capillary gas chromatography (GC).
Catalog No. 50-211-2910