Learn More
Medchemexpress LLC HY-112929 5mg Medchemexpress, DT-061 CAS:1809427-19-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1129295MG
Encompass_Preferred
Medchemexpress, HY-112929 5mg DT-061 CAS:1809427-19-7 DT-061 is an orally bioavailable activator of protein phosphatase 2A (PP2A) and could be applied in the therapy of KRAS-mutant and MYC-driven tumorigenesis [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2445