Learn More
Selleck Chemical LLC DT-061 (SMAP) 5mg 1809427-19-7
SDP
Supplier:  Selleck Chemical LLC S87745MG
Encompass_Preferred
DT-061 (SMAP) is a bioavailable PP2A activator. It decreases cell viability in HCC827 and HCC3255 cell lines, with IC50 values of 14.3 ?M and 12.4 ?M respectively. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC1918492