Learn More
Medchemexpress LLC DT-3 acetate | 98.9% | 3294.07 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYP0142A10MG
Encompass_Preferred
DT-3 acetate is a membrane-permeable protein kinase G Iα (PKG Iα) inhibitory peptide that pharmacologically blocks cGMP-PKG signaling.
  • Acts as a PKG Iα inhibitory peptide
  • Pharmacologically blocks cGMP-PKG signaling
  • Suitable for research in blood or cardio-cerebrovascular disease
  • Useful in cardiovascular disease studies
  • High purity of 98.9%
  • Soluble in H2O at ≥ 100 mg/mL
Catalog No. 50-004-25936