Learn More
Bia Separartions Inc DT18 4ML CGMP COMPL MONO CLM
Supplier:  Bia Separartions Inc BIA904.12192
Encompass
CIMmultus™ Oligo dT18 4 mL cGMP Compliant Monolithic Column (C12 Linker) (2 μm channels)
Catalog No. NC2122736
Add to Cart
Request a Quote