Learn More
Selleck Chemical LLC DT2216-1mg
SDP
Supplier:  Selleck Chemical LLC S89241MG
Encompass_Preferred
DT2216 is a potent and selective degrader of BCL-XL based on PROTAC technology. DT2216 inhibits various BCL-XL-dependent leukemia and cancer cells but considerably less toxic to platelets.
Catalog No. 50-238-9705